accessibility menu, dialog, popup

UserName

CAS RN 121-63-1

ClSOOOSClOO

CAS RN 121-63-1

IUPAC Name: 4-[4-(chlorosulfonyl)phenoxy]benzene-1-sulfonyl chloride
Synonyms: 4-[4-(chlorosulfonyl)phenoxy]benzenesulfonyl chloride
Molecular Weight (g/mol): 367.21
Molecular Formula: C12H8Cl2O5S2
InChi Key: HJKXLQIPODSWMB-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(OC2=CC=C(C=C2)S(Cl)(=O)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results