accessibility menu, dialog, popup

UserName

CAS RN 1208-64-6

CH3H3CCH3OO

CAS RN 1208-64-6

IUPAC Name: 3-(4-tert-butylphenyl)propanoate
Synonyms: 3-(4-tert-butylphenyl)propanoate
Molecular Weight (g/mol): 205.28
Molecular Formula: C13H17O2
InChi Key: BNJYANVQFVSYEK-UHFFFAOYSA-M
SMILES: CC(C)(C)C1=CC=C(CCC([O-])=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results