accessibility menu, dialog, popup

UserName

CAS RN 12073-36-8

PtPtClClClCl

CAS RN 12073-36-8

IUPAC Name: bis(dichloroplatinum); bis(ethene)
Synonyms: bis(ethylene); bis(platinum(II) chloride)
Molecular Weight (g/mol): 588.08
Molecular Formula: C4H8Cl4Pt2
InChi Key: XBLBDYCZFZESPN-UHFFFAOYSA-J
SMILES: C=C.C=C.Cl[Pt]Cl.Cl[Pt]Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results