accessibility menu, dialog, popup

UserName

CAS RN 12069-32-8

BBBB

CAS RN 12069-32-8

IUPAC Name: 2,3,4,5-tetraboratetracyclo[2.1.0.0¹,³.0²,⁵]pentane
Synonyms: boron carbide
Molecular Weight (g/mol): 55.25
Molecular Formula: CB4
InChi Key: INAHAJYZKVIDIZ-UHFFFAOYSA-N
SMILES: B12B3B4B1C234
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)