accessibility menu, dialog, popup

UserName

CAS RN 1204501-38-1

H3COONNCl

CAS RN 1204501-38-1

IUPAC Name: ethyl 7-chloroimidazo[1,2-a]pyridine-2-carboxylate
Synonyms: ethyl 7-chloroimidazo[1,2-a]pyridine-2-carboxylate
Molecular Weight (g/mol): 224.64
Molecular Formula: C10H9ClN2O2
InChi Key: ZGSKNRFGZMYUGO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN2C=CC(Cl)=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results