accessibility menu, dialog, popup

UserName

CAS RN 12045-64-6

IUPAC Name: diboron zirconium
Synonyms: zirconium diboride
Molecular Weight (g/mol): 112.84
Molecular Formula: B2Zr
InChi Key: VWZIXVXBCBBRGP-UHFFFAOYSA-N
SMILES: [B].[B].[Zr]
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results