accessibility menu, dialog, popup

UserName

CAS RN 12004-39-6

OOOOOOOAlAlTiTi

CAS RN 12004-39-6

IUPAC Name: dititanium(4+) dialuminium(3+) heptaoxidandiide
Synonyms: dititanium(4+) dialuminium(3+) heptaoxidandiide
Molecular Weight (g/mol): 261.69
Molecular Formula: Al2O7Ti2
InChi Key: COTUMVFAJGQVPQ-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[O--].[O--].[Al+3].[Al+3].[Ti+4].[Ti+4]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results