CAS RN 1198-37-4
CAS RN 1198-37-4
IUPAC Name:
2,4-dimethylquinoline
Synonyms:
2,4-dimethylquinoline
Molecular Weight (g/mol):
157.22
Molecular Formula:
C11H11N
InChi Key:
ZTNANFDSJRRZRJ-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=C2C=CC=CC2=N1
1
–
4
of
4
results
1
2,4-Dimethylquinoline, 95%
CAS: 1198-37-4 Molecular Formula: C11H11N Molecular Weight (g/mol): 157.216 MDL Number: MFCD00006760 InChI Key: ZTNANFDSJRRZRJ-UHFFFAOYSA-N PubChem CID: 14536 IUPAC Name: 2,4-dimethylquinoline SMILES: CC1=CC(=NC2=CC=CC=C12)C
2
2,4-Dimethylquinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,4-Dimethylquinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
2,4-Dimethylquinoline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.