accessibility menu, dialog, popup

UserName

CAS RN 1196147-72-4

H3CCH3H3CPNH2CH3CH3H3C

CAS RN 1196147-72-4

IUPAC Name: (3-aminopropyl)di-tert-butyl)phosphane
Synonyms: (3-aminopropyl)di-tert-butyl)phosphane
Molecular Weight (g/mol): 203.31
Molecular Formula: C11H26NP
InChi Key: PREMCOLELPAPBB-UHFFFAOYSA-N
SMILES: CC(C)(C)P(CCCN)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)