CAS RN 1190021-64-7
CAS RN 1190021-64-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Naratriptan-d3 Hydrochloride, TRC
CAS: 1190021-64-7 Molecular Formula: C17H23D3ClN3O2S Molecular Weight (g/mol): 374.94 Synonym: N-Methyl-d3-2-[3-(1-methyl-4-piperidyl)-1H-indol-5-yl]-ethanesulfonamide Hydrochloride,Naramig-d3,Amerge-d3. SMILES: O=S(CCC1=CC=C2C(C(C3CCN(C([2H])([2H])[2H])CC3)=CN2)=C1)(NC)=O.Cl