CAS RN 1189654-25-8
CAS RN 1189654-25-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Ethyl (5-Ethyl-2-pyridinyl)-1,1-d2-acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Ethyl (5-Ethyl-2-pyridinyl)-1,1-d2-acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.