accessibility menu, dialog, popup

UserName

CAS RN 1184-16-3

N(R)(R)(R)NN(R)OOPPPO(R)(R)(R)(S)ONNOOOHOOHOOOOHOONH2NH2HOOHNa

CAS RN 1184-16-3

IUPAC Name: 1-{5-[({[({[5-(6-amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)oxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)methyl]-3,4-dihydroxyoxolan-2-yl}-3-carbamoyl-1λ⁵-pyridin-1-ylium
Synonyms: 1-(5-{[({[5-(6-aminopurin-9-yl)-3-hydroxy-4-(phosphonooxy)oxolan-2-yl]methoxy(hydroxy)phosphoryl}oxy(hydroxy)phosphoryl)oxy]methyl}-3,4-dihydroxyoxolan-2-yl)-3-carbamoyl-1λ⁵-pyridin-1-ylium
Molecular Weight (g/mol): 744.42
Molecular Formula: C21H29N7O17P3
InChi Key: XJLXINKUBYWONI-UHFFFAOYNA-O
SMILES: NC(=O)C1=CC=C[N+](=C1)C1OC(COP(O)(=O)OP(O)(=O)OCC2OC(C(OP(O)(O)=O)C2O)N2C=NC3=C(N)N=CN=C23)C(O)C1O
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results