CAS RN 118156-93-7
CAS RN 118156-93-7
IUPAC Name:
tert-butyl 3-formylpiperidine-1-carboxylate
Synonyms:
tert-butyl 3-formylpiperidine-1-carboxylate
Molecular Weight (g/mol):
213.28
Molecular Formula:
C11H19NO3
InChi Key:
CTVHINDANRPFIL-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)N1CCCC(C1)C=O
1
–
1
of
1
results
1
1-Boc-piperidine-3-carboxaldehyde, 97%
CAS: 118156-93-7 Molecular Formula: C11H19NO3 Molecular Weight (g/mol): 213.277 MDL Number: MFCD02179020 InChI Key: CTVHINDANRPFIL-UHFFFAOYSA-N Synonym: 1-boc-3-piperidinecarboxaldehyde,boc-3-piperidinecarboxaldehyde,3-formylpiperidine-1-carboxylic acid tert-butyl ester,n-boc-3-formylpiperidine,1-boc-3-formylpiperidine,1-boc-piperidine-3-carboxaldehyde,3-formyl-n-tert-butoxycarbonyl piperidine,1-piperidinecarboxylic acid, 3-formyl-, 1,1-dimethylethyl ester,n-boc-3-piperidine carboxyaldehyde,1-n-boc-3-formylpiperidine PubChem CID: 9964448 IUPAC Name: tert-butyl 3-formylpiperidine-1-carboxylate SMILES: CC(C)(C)OC(=O)N1CCCC(C1)C=O