CAS RN 1175018-73-1
CAS RN 1175018-73-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Despropargyl N-(2-Chloroallyl) Rasagiline, TRC
CAS: 1175018-73-1 Molecular Formula: C12 H14 Cl N Molecular Weight (g/mol): 207.7 Synonym: N-Despropargyl N-(2-Chloroallyl) Rasagiline,(1R)-N-(2-Chloro-2-propen-1-yl)-2,3-dihydro-1H-inden-1-amine IUPAC Name: (1R)-N-(2-chloroallyl)indan-1-amine SMILES: ClC(=C)CN[C@@H]1CCc2ccccc12