CAS RN 1174289-22-5
CAS RN 1174289-22-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bazedoxifene N-Oxide, TRC
CAS: 1174289-22-5 Molecular Formula: C30 H34 N2 O4 Molecular Weight (g/mol): 486.6 Synonym: 1H-Indol-5-ol, 1-[[4-[2-(hexahydro-1-oxido-1H-azepin-1-yl)ethoxy]phenyl]methyl]-2-(4-hydroxyphenyl)-3-methyl-,1-[[4-[2-(Hexahydro-1-oxido-1H-azepin-1-yl)ethoxy]phenyl]methyl]-2-(4-hydroxyphenyl)-3-methyl-1H-indol-5-ol,Bazedoxifene N-Oxide IUPAC Name: 2-(4-hydroxyphenyl)-3-methyl-1-[[4-[2-(1-oxidoazepan-1-ium-1-yl)ethoxy]phenyl]methyl]indol-5-ol SMILES: Cc1c(c2ccc(O)cc2)n(Cc3ccc(OCC[N+]4([O-])CCCCCC4)cc3)c5ccc(O)cc15