accessibility menu, dialog, popup

UserName

CAS RN 116834-64-1

SOONHOHON

CAS RN 116834-64-1

IUPAC Name: 4-(quinoline-8-sulfonamido)benzoic acid
Synonyms: 4-(quinoline-8-sulfonamido)benzoic acid
Molecular Weight (g/mol): 328.34
Molecular Formula: C16H12N2O4S
InChi Key: NBQVTLMGAPBCJA-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(NS(=O)(=O)C2=C3N=CC=CC3=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results