CAS RN 116661-27-9
CAS RN 116661-27-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-Haloxyfop-methyl, TRC
CAS: 116661-27-9 Molecular Formula: C17 H19 N O4 Molecular Weight (g/mol): 301.337 Synonym: (S)-2-[4-[[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy]phenoxy]-propanoic Acid Methyl Ester; IUPAC Name: (4R,4aR,7S,7aR,12bS)-3-methyl-3-oxo-2,4,4a,7,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7,9-diol SMILES: CN1(=O)CC[C@]23[C@H]4Oc5c(O)ccc(C[C@@H]1[C@@H]2C=C[C@@H]4O)c35