accessibility menu, dialog, popup

UserName

CAS RN 1164-16-5

(S)OHNOHHOOO

CAS RN 1164-16-5

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
Molecular Weight (g/mol): 315.33
Molecular Formula: C17H17NO5
InChi Key: MCRMUCXATQAAMN-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)