accessibility menu, dialog, popup

UserName

CAS RN 116183-81-4

HNNH2HClHCl

CAS RN 116183-81-4

IUPAC Name: pyrrolidin-3-amine dihydrochloride
Synonyms: pyrrolidin-3-amine dihydrochloride
Molecular Weight (g/mol): 159.05
Molecular Formula: C4H12Cl2N2
InChi Key: NJPNCMOUEXEGBL-UHFFFAOYNA-N
SMILES: Cl.Cl.NC1CCNC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)