accessibility menu, dialog, popup

UserName

CAS RN 115932-00-8

H3COONNN

CAS RN 115932-00-8

IUPAC Name: ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate
Synonyms: ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate
Molecular Weight (g/mol): 191.19
Molecular Formula: C9H9N3O2
InChi Key: JEMAMNBFHPIPCR-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C2N=CC=CN2N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results