accessibility menu, dialog, popup

UserName

CH3OOOCH3O

CAS RN 1157-39-7

IUPAC Name: 7-methoxy-3-(4-methoxyphenyl)-4H-chromen-4-one
Synonyms: 7,4'-di-O-methyldaidzein
Molecular Weight (g/mol): 282.3
Molecular Formula: C17H14O4
InChi Key: LPNBCGIVZXHHHO-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=COC2=CC(OC)=CC=C2C1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results