accessibility menu, dialog, popup

UserName

CAS RN 1154-59-2

OHClClONHClCl

CAS RN 1154-59-2

IUPAC Name: 3,5-dichloro-N-(3,4-dichlorophenyl)-2-hydroxybenzamide
Synonyms: Tcsa TCSA
Molecular Weight (g/mol): 351
Molecular Formula: C13H7Cl4NO2
InChi Key: SJQBHPJLLIJASD-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=C(Cl)C=C1C(=O)NC1=CC=C(Cl)C(Cl)=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results