accessibility menu, dialog, popup

UserName

CAS RN 115338-32-4

HBrH3CONNNOO

CAS RN 115338-32-4

IUPAC Name: hydrogen 2-{4-[4-(2-methoxyphenyl)piperazin-1-yl]butyl}-2,3-dihydro-1H-isoindole-1,3-dione bromide
Synonyms: hydrogen 2-{4-[4-(2-methoxyphenyl)piperazin-1-yl]butyl}isoindole-1,3-dione bromide
Molecular Weight (g/mol): 474.4
Molecular Formula: C23H28BrN3O3
InChi Key: AXRUEPFPTQYHQD-UHFFFAOYSA-N
SMILES: [H+].[Br-].COC1=CC=CC=C1N1CCN(CCCCN2C(=O)C3=CC=CC=C3C2=O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results