accessibility menu, dialog, popup

UserName

H3CNH3CCH3ClICl

CAS RN 114971-52-7

IUPAC Name: benzyltrimethylazanium; dichloroiodanuide
Synonyms: benzyltrimethylazanium; dichloroiodanuide
Molecular Weight (g/mol): 348.05
Molecular Formula: C10H16Cl2IN
InChi Key: PPDJNZTUDFPAHX-UHFFFAOYSA-N
SMILES: Cl[I-]Cl.C[N+](C)(C)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)