accessibility menu, dialog, popup

UserName

CAS RN 1149-26-4

H3C(S)OHHNCH3OOO

CAS RN 1149-26-4

IUPAC Name: (2R)-2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoate
Synonyms: (2R)-2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoate
Molecular Weight (g/mol): 250.28
Molecular Formula: C13H16NO4
InChi Key: CANZBRDGRHNSGZ-LLVKDONJSA-M
SMILES: CC(C)[C@@H](NC(=O)OCC1=CC=CC=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)