accessibility menu, dialog, popup

UserName

CH3OHNN

CAS RN 114636-30-5

IUPAC Name: N-(1-benzylpyrrolidin-3-yl)acetamide
Synonyms: N-(1-benzylpyrrolidin-3-yl)acetamide
Molecular Weight (g/mol): 218.3
Molecular Formula: C13H18N2O
InChi Key: CMSWETNAAPYFSH-UHFFFAOYNA-N
SMILES: CC(=O)NC1CCN(CC2=CC=CC=C2)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results