CAS RN 1140251-30-4
CAS RN 1140251-30-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-[4-Amino-2-chloro-6-[(2-deoxy-2-fluoro-alpha-D-arabinopyranosyl)amino]-5-pyrimidinyl]-formamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-[4-Amino-2-chloro-6-[(2-deoxy-2-fluoro-Alpha-D-arabinopyranosyl)amino]-5-pyrimidinyl]-formamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.