CAS RN 113713-60-3
IUPAC Name:
5-methoxy-1H,2H,3H-imidazo[4,5-b]pyridine-2-thione
Synonyms:
5-methoxy-1H,3H-imidazo[4,5-b]pyridine-2-thione
Molecular Weight (g/mol):
181.21
Molecular Formula:
C7H7N3OS
InChi Key:
PNYFSMIUARCIRR-UHFFFAOYSA-N
SMILES:
COC1=CC=C2NC(=S)NC2=N1
1
–
1
of
1
results
1
2-Mercapto-5-methoxy-3H-imidazo[4,5-b]pyridine, 95%, Thermo Scientific™
CAS: 113713-60-3 Molecular Formula: C7H7N3OS Molecular Weight (g/mol): 181.213 MDL Number: MFCD06658230 InChI Key: PNYFSMIUARCIRR-UHFFFAOYSA-N Synonym: 2-mercapto-5-methoxyimidazole 4,5-b pyridine,2-mercapto-5-methoxyimidazole-4,5-b pyridine,5-methoxy-3h-imidazo 4,5-b pyridine-2-thiol,5-methoxy-1h-imidazo 4,5-b pyridine-2-thiol,2-mercapto-5-methoxy-3h-imidazo 4,5-b pyridine,2-mercapto-5-methoxyimidazol 4,5-b pyridine,5-methoxy-1,3-dihydro-2h-imidazo 4,5-b pyridine-2-thione,2h-imidazo 4,5-b pyridine-2-thione, 1,3-dihydro-5-methoxy,pubchem15037 PubChem CID: 11126879 IUPAC Name: 5-methoxy-1,3-dihydroimidazo[4,5-b]pyridine-2-thione SMILES: COC1=NC2=C(C=C1)NC(=S)N2