CAS RN 1135366-22-1
CAS RN 1135366-22-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(2R)-2-Hydroxy-4-[(4-methoxyphenyl)methoxy]-N,N,N-trimethyl-4-oxo-1-butanaminium Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2R)-2-Hydroxy-4-[(4-methoxyphenyl)methoxy]-N,N,N-trimethyl-4-oxo-1-butanaminium Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(2R)-2-Hydroxy-4-[(4-methoxyphenyl)methoxy]-N,N,N-trimethyl-4-oxo-1-butanaminium Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.