accessibility menu, dialog, popup

UserName

CAS RN 1135280-78-2

NOFNONHSHCl

CAS RN 1135280-78-2

IUPAC Name: 3-{2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl}-2-sulfanylidene-1,2,3,4-tetrahydroquinazolin-4-one hydrochloride
Synonyms: altanserin hydrochloride
Molecular Weight (g/mol): 447.95
Molecular Formula: C22H23ClFN3O2S
InChi Key: JFPPLMAMMZZOEA-UHFFFAOYSA-N
SMILES: Cl.FC1=CC=C(C=C1)C(=O)C1CCN(CCN2C(=S)NC3=CC=CC=C3C2=O)CC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results