accessibility menu, dialog, popup

UserName

CAS RN 1129-69-7

H3CN

CAS RN 1129-69-7

IUPAC Name: 2-hexylpyridine
Synonyms: pyridine, 2-hexyl-
Molecular Weight (g/mol): 163.26
Molecular Formula: C11H17N
InChi Key: NZLJDTKLZIMONR-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results