CAS RN 112093-28-4
CAS RN 112093-28-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(Z)-4-Hydroxy-N-desmethyl Tamoxifen (contains up to 10% E isomer), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(Z)-4-Hydroxy-N-desmethyl Tamoxifen (contains up to 10% E isomer), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(Z)-4-Hydroxy-N-desmethyl Tamoxifen (contains up to 10% E isomer), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.