accessibility menu, dialog, popup

UserName

CAS RN 1120-73-6

CH3O

CAS RN 1120-73-6

IUPAC Name: 2-methylcyclopent-2-en-1-one
Synonyms: 2-cyclopenten-1-one, 2-methyl-
Molecular Weight (g/mol): 96.13
Molecular Formula: C6H8O
InChi Key: ZSBWUNDRDHVNJL-UHFFFAOYSA-N
SMILES: CC1=CCCC1=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results