accessibility menu, dialog, popup

UserName

CAS RN 1120-01-0

NaH3CSOOO

CAS RN 1120-01-0

IUPAC Name: sodium hexadecane-1-sulfonate
Synonyms: sodium hexadecane-1-sulphonate
Molecular Weight (g/mol): 328.49
Molecular Formula: C16H33NaO3S
InChi Key: PNGBYKXZVCIZRN-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCCCCCCCCS([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results