accessibility menu, dialog, popup

UserName

CAS RN 111-43-3

H3COCH3

CAS RN 111-43-3

IUPAC Name: 1-propoxypropane
Synonyms: DI(N-propyl) ether
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: POLCUAVZOMRGSN-UHFFFAOYSA-N
SMILES: CCCOCCC
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)