accessibility menu, dialog, popup

UserName

NFClClFFF

CAS RN 1099598-11-4

IUPAC Name: 2,6-dichloro-3-fluoro-5-(trifluoromethyl)pyridine
Synonyms: 2,6-dichloro-3-fluoro-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 233.97
Molecular Formula: C6HCl2F4N
InChi Key: VVXQCTFINCODIF-UHFFFAOYSA-N
SMILES: FC1=C(Cl)N=C(Cl)C(=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results