accessibility menu, dialog, popup

UserName

ClSO

CAS RN 107572-07-6

IUPAC Name: 2-[(4-chlorophenyl)sulfanyl]benzaldehyde
Synonyms: 2-[(4-chlorophenyl)sulfanyl]benzaldehyde
Molecular Weight (g/mol): 248.72
Molecular Formula: C13H9ClOS
InChi Key: CQKLAEUCMKGSEQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(SC2=CC=CC=C2C=O)C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)