accessibility menu, dialog, popup

UserName

CH3NOOHO

CAS RN 107367-98-6

IUPAC Name: 2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid
Synonyms: (5-methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid
Molecular Weight (g/mol): 217.22
Molecular Formula: C12H11NO3
InChi Key: XEWJNPORMBGGKZ-UHFFFAOYSA-N
SMILES: CC1=C(CC(O)=O)N=C(O1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results