CAS RN 1073371-90-0
CAS RN 1073371-90-0
IUPAC Name:
2-(cyclopentyloxy)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms:
2-(cyclopentyloxy)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol):
289.18
Molecular Formula:
C16H24BNO3
InChi Key:
GZCZGFSEHAZEPS-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=C(OC2CCCC2)N=CC=C1
1
–
1
of
1
results
1
2-Cyclopentyloxypyridine-3-boronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 1073371-90-0 Molecular Formula: C16H24BNO3 Molecular Weight (g/mol): 289.18 MDL Number: MFCD07781170 InChI Key: GZCZGFSEHAZEPS-UHFFFAOYSA-N Synonym: 2-cyclopentyloxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,2-cyclopentyloxypyridine-3-boronic acid pinacol ester,2-cyclopentyloxy pyridine-3-boronic acid pinacol ester,2-cyclopentyloxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,2-cyclopentyloxy pyridine-3-boronic acid, pinacol ester,2-cyclopentyloxy pyridin-3-yl boronic acid pinacol ester PubChem CID: 24208792 IUPAC Name: 2-cyclopentyloxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine SMILES: CC1(C)OB(OC1(C)C)C1=C(OC2CCCC2)N=CC=C1