CAS RN 107254-86-4
CAS RN 107254-86-4
IUPAC Name:
5-nitro-2-[(3-phenylpropyl)amino]benzoic acid
Synonyms:
NPPB
Molecular Weight (g/mol):
300.31
Molecular Formula:
C16H16N2O4
InChi Key:
WBSMIPAMAXNXFS-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(=CC=C1NCCCC1=CC=CC=C1)[N+]([O-])=O