CAS RN 1070-10-6
CAS RN 1070-10-6
IUPAC Name:
titanium(4+) tetrakis(2-ethylhexan-1-olate)
Synonyms:
titanium(4+) tetrakis(2-ethylhexan-1-olate)
Molecular Weight (g/mol):
564.76
Molecular Formula:
C32H68O4Ti
InChi Key:
KTXWGMUMDPYXNN-UHFFFAOYNA-N
SMILES:
[Ti+4].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-]
1
–
1
of
1
results
1
Titanium(IV) 2-ethylhexyloxide, 95%, Thermo Scientific™
CAS: 1070-10-6 Molecular Formula: C32H68O4Ti Molecular Weight (g/mol): 564.76 MDL Number: MFCD00015264,MFCD00015264 InChI Key: KTXWGMUMDPYXNN-UHFFFAOYNA-N Synonym: tyzor tot,2-ethylhexyl orthotitanate,tetra 2-ethylhexyl titanate,tetra-2-ethylhexyl titanate,tetrakis 2-ethylhexyl titanate,titanic iv acid, tetrakis 2-ethylhexyl ester,1-hexanol, titanium 4+ salt PubChem CID: 54603245 IUPAC Name: titanium(4+) tetrakis(2-ethylhexan-1-olate) SMILES: [Ti+4].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-]