CAS RN 107-85-7
CAS RN 107-85-7
IUPAC Name:
3-methylbutan-1-amine
Synonyms:
isoamylamine leucamine isobutylcarbylamine 1-aminoisopentane
Molecular Weight (g/mol):
87.17
Molecular Formula:
C5H13N
InChi Key:
BMFVGAAISNGQNM-UHFFFAOYSA-N
SMILES:
CC(C)CCN
1
–
2
of
2
results
2
Isoamylamine, 99%, Thermo Scientific Chemicals
CAS: 107-85-7 Molecular Formula: C5H13N Molecular Weight (g/mol): 87.15 MDL Number: MFCD00008203 InChI Key: BMFVGAAISNGQNM-UHFFFAOYSA-N Synonym: isoamylamine,isopentylamine,3-methylbutylamine,1-amino-3-methylbutane,monoisoamylamine,leucamine,1-butanamine, 3-methyl,3-methylbutanamine,isovalerylamine,monoisopentylamine PubChem CID: 7894 ChEBI: CHEBI:43689 IUPAC Name: 3-methylbutan-1-amine SMILES: CC(C)CCN