CAS RN 107-70-0
CAS RN 107-70-0
IUPAC Name:
4-methoxy-4-methylpentan-2-one
Synonyms:
4-methoxy-4-methyl-2-pentanone
Molecular Weight (g/mol):
130.19
Molecular Formula:
C7H14O2
InChi Key:
KOKPBCHLPVDQTK-UHFFFAOYSA-N
SMILES:
COC(C)(C)CC(C)=O
1
–
1
of
1
results
1
4-Methoxy-4-methyl-2-pentanone, 97%, Thermo Scientific Chemicals
CAS: 107-70-0 Molecular Formula: C7H14O2 Molecular Weight (g/mol): 130.187 MDL Number: MFCD00043932 InChI Key: KOKPBCHLPVDQTK-UHFFFAOYSA-N Synonym: 4-methoxy-4-methyl-2-pentanone,pentoxone,pent-oxone solvent,2-pentanone, 4-methoxy-4-methyl,4-methyl-4-methoxy-2-pentanone,me 6 k,ccris 4840,acmc-1bwvk,dsstox_cid_5557,dsstox_rid_77828 PubChem CID: 7884 IUPAC Name: 4-methoxy-4-methylpentan-2-one SMILES: CC(=O)CC(C)(C)OC