accessibility menu, dialog, popup

UserName

CAS RN 106876-54-4

IUPAC Name: 2,2-difluoro-2H-1,3-benzodioxol-4-amine
Synonyms: 2,2-difluoro-1,3-benzodioxol-4-amine
Molecular Weight (g/mol): 173.12
Molecular Formula: C7H5F2NO2
InChi Key: RXIKYXZKSIARLN-UHFFFAOYSA-N
SMILES: NC1=CC=CC2=C1OC(F)(F)O2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results