CAS RN 106447-97-6
CAS RN 106447-97-6
IUPAC Name:
4-(trifluoromethyl)pyridin-2-amine
Synonyms:
4-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol):
162.12
Molecular Formula:
C6H5F3N2
InChi Key:
RWGBXAQMUBGGKQ-UHFFFAOYSA-N
SMILES:
NC1=NC=CC(=C1)C(F)(F)F
1
–
1
of
1
results
1
2-Amino-4-(trifluoromethyl)pyridine, 99%, Thermo Scientific Chemicals
CAS: 106447-97-6 Molecular Formula: C6H5F3N2 Molecular Weight (g/mol): 162.115 MDL Number: MFCD00077455 InChI Key: RWGBXAQMUBGGKQ-UHFFFAOYSA-N Synonym: 2-amino-4-trifluoromethyl pyridine,4-trifluoromethyl pyridin-2-amine,4-trifluoromethyl-pyridin-2-ylamine,2-amino-4-trifluoromethylpyridine,4-trifluoromethyl-2-pyridinamine,4-trifluoromethyl-2-pyridylamine,2-pyridinamine, 4-trifluoromethyl,2-amino-4-trifluoromethyl-pyridine,pubchem2977 PubChem CID: 2778315 IUPAC Name: 4-(trifluoromethyl)pyridin-2-amine SMILES: C1=CN=C(C=C1C(F)(F)F)N