accessibility menu, dialog, popup

Test

CAS RN 105752-11-2

CAS RN 105752-11-2

IUPAC Name: 4-iodopyridin-3-amine
Synonyms: 4-iodopyridin-3-amine
Molecular Weight (g/mol): 220.01
Molecular Formula: C5H5IN2
InChi Key: ZJRSKTXMSIVNAU-UHFFFAOYSA-N
SMILES: NC1=CN=CC=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results