CAS RN 104748-87-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(3-Chloro-6,11-dihydro-6-methyl-5,5-dioxidodibenzo[c,f][1,2]thiazepin-11-yl)-Beta-alanine, TRC
CAS: 104748-87-0 Molecular Formula: C17H17ClN2O4S Molecular Weight (g/mol): 380.85 Synonym: Tianeptine Metabolite MC3; SMILES: CN1c2ccccc2C(NCCC(=O)O)c3ccc(Cl)cc3S1(=O)=O