CAS RN 10392-74-2
CAS RN 10392-74-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
9-Chloro Triamcinolone Acetonide, TRC
CAS: 10392-74-2 Molecular Formula: C24 H31 Cl O6 Molecular Weight (g/mol): 450.95 Synonym: Pregna-1,4-diene-3,20-dione, 9-chloro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]-, (11β,16α)- (9CI),Pregna-1,4-diene-3,20-dione, 9-chloro-11β,16α,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone (7CI,8CI),2H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole, pregna-1,4-diene-3,20-dione deriv. IUPAC Name: (1S,2S,4R,8S,9S,11S,12R,13S)-12-chloro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-16-one SMILES: CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(Cl)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO