CAS RN 10362-82-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
alpha-Methylstyrene-d10, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
Alpha-Methylstyrene-d10, TRC
CAS: 10362-82-0 Molecular Formula: C9 D10 Molecular Weight (g/mol): 128.237 Synonym: Styrene-β,β,2,3,4,5,6-d7, α-methyl-d3- (8CI),α-(Methyl-d3)styrene-β,β,2,3,4,5,6-d7,[1-(Methyl-d3)ethenyl-2,2-d2]benzene-2,3,4,5,6-d5,α-Methylstyrene-d10,α-Methylstyrene-D10 IUPAC Name: 1,2,3,4,5-pentadeuterio-6-(1,1,3,3,3-pentadeuterioprop-1-en-2-yl)benzene SMILES: [2H]C(=C(c1c([2H])c([2H])c([2H])c([2H])c1[2H])C([2H])([2H])[2H])[2H]