CAS RN 103335-49-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
S-2-Pyridyl 3-oxo-4-aza-5a-androst-1-ene-17b-carbothioate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
S-2-Pyridyl 3-oxo-4-aza-5Alpha-androst-1-ene-17Beta-carbothioate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
S-2-Pyridyl 3-oxo-4-aza-5Alpha-androst-1-ene-17Beta-carbothioate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.